Invitrogen
Manufacturer Code:PA547237
Catalog # PIPA547237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Endocan/ESM-1 Trp20-Arg184 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Endothelial cell-specific molecule 1; ESM-1; ESM1 ESM-1 IGFBP-rp6 endocan endothelial cell-specific molecule 1 ESM-1
Gene Aliases: 0610042H23Rik; AV004503; ESM-1; Esm1
UniProt ID: (Mouse) Q9QYY7
Entrez Gene ID: (Mouse) 71690
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.