Invitrogen
Manufacturer Code:PA520004
Catalog # PIPA520004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids near the amino terminus of human EndoG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endo G; endo G endonuclease G mitochondrial mitochondrial endonuclease G; Endonuclease G, mitochondrial; mitochondrial endonuclease G
Gene Aliases: ENDOG
UniProt ID: (Human) Q14249, (Mouse) O08600
Entrez Gene ID: (Human) 2021, (Mouse) 13804, (Rat) 362100
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.