Novus Biologicals
Manufacturer Code:NBP179222
Catalog # NBP179222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human EMX1The immunogen for this antibody is EMX1. Peptide sequence DGLLLHGPFARKPKRIRTAFSPSQLLRLERAFEKNHYVVGAERKQLAGSL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: empty spiracles homeobox 1 empty spiracles-like protein 1; Empty spiracles homolog 1; Empty spiracles-like protein 1; Homeobox protein EMX1
Gene Aliases: EMX1
UniProt ID: (Human) Q04741
Entrez Gene ID: (Human) 2016
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.