Invitrogen
Manufacturer Code:PA523257
Catalog # PIPA523257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (143 FFREEKEQRGTFNFKVPE 160) of Embigin protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Embigin; embigin homolog; glycoprotein 70; Teratocarcinoma glycoprotein Gp-70
Gene Aliases: AL022799; EMB; GP70
UniProt ID: (Human) Q6PCB8, (Mouse) P21995
Entrez Gene ID: (Human) 133418, (Mouse) 13723
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.