Novus Biologicals
Manufacturer Code:NBP243694
Catalog # NBP243694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11811 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ELP3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1.48 elongation protein 3 homolog (S. cerevisiae) elongator complex protein 3 hELP3 KAT9FLJ10422; elongation protein 3 homolog; Elongator complex protein 3; hELP3; Protein lysine acetyltransferase ELP3; tRNA uridine(34) acetyltransferase
Gene Aliases: ELP3; KAT9
UniProt ID: (Human) Q9H9T3
Entrez Gene ID: (Human) 55140
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.