Abnova
Manufacturer Code:PAB14063
Catalog # 89-116-904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of synthetic peptides (conjugated with KLH) that corresponding to internal region of mouse Ehmt2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ehmt2.
For Research Use Only
Protein Aliases: Bat8; D17Ertd710e; Euchromatic histone-lysine N-methyltransferase 2; G9a; H3-K9-HMTase 3; Histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B-associated transcript 8; KMT1C; NG36; Protein G9a
Gene Aliases: Bat8; D17Ertd710e; Ehmt2; G9a; KMT1C; NG36
UniProt ID: (Mouse) Q9Z148
Entrez Gene ID: (Mouse) 110147
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.