Invitrogen
Manufacturer Code:12669180
Catalog # 50-245-517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | erongr2 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; Early growth response protein 2; EGR-2; KROX-20 early growth response gene 2 CMT1D CMT4E DKFZp686J1957 FLJ1454; Zinc finger protein Krox-20
Gene Aliases: Egr-2; Egr2; Krox-20; Krox20; NGF1-B; Zfp-25; Zfp-6
UniProt ID: (Mouse) P08152
Entrez Gene ID: (Mouse) 13654
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.