Novus Biologicals
Manufacturer Code:NBP187957
Catalog # NBP187957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ERAPENCHLANEIKPSDPPLDNQMKHSFDSASNKNFSQCLESKLENSPVENVTAASTLLSQAKIDTGENKFPGSAPQQHSILSNQTSKSSDNRETPRNHSLPKCNSHLEI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTF7 homolog 1; CTF7 homolog 1 EC 2.3.1.- ECO1 homolog 1 EFO1CTF EFO1p ESO1 homolog 1 ESO1A930014I12Rik Establishment factor-like protein 1 Establishment of cohesion 1 homolog 1 establishment of cohesion 1 homolog 1 (S. cerevisiae) hEFO1 KIAA1911ECO1 MGC105022 N-acetyltransferase ESCO1; ECO1 homolog 1; EFO1; EFO1p; ESO1 homolog 1; Establishment factor-like protein 1; Establishment of cohesion 1 homolog 1; hEFO1; N-acetyltransferase ESCO1; N-acetyltransferase ESCO1 variant 2
Gene Aliases: A930014I12Rik; CTF; ECO1; EFO1; ESCO1; ESO1; KIAA1911
UniProt ID: (Human) Q5FWF5
Entrez Gene ID: (Human) 114799
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.