Novus Biologicals
Manufacturer Code:NB10062537
Catalog # NB10062537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the amino acids 78 - 92 of the native molecule conjugated to Keyhole limpet Haemocyanin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 20-hydroxy-ecdysone receptor; 20E receptor; 20E receptor EcR EcRH NR1H1; CG1765-PA; CG1765-PB; CG1765-PC; CG1765-PD; CG1765-PE; CG1765-PG; ecdyson receptor 1b; Ecdysone receptor; ecdysone receptor complex; Ecdysone-R; ecdysone-Receptor; Ecdysteroid receptor; ecdysterone receptor; EcR-PA; EcR-PB; EcR-PC; EcR-PD; EcR-PE; EcR-PG; EcRH; long island expressway; male sterile(2)42A; Nuclear receptor subfamily 1 group H member 1; RE06878p; RE33854p; snaggletooth
Gene Aliases: anon-WO0229075.1; CG1765; CG8347; DEcR; Dhr23; DmEcR; Dmel\CG1765; Dmel_CG1765; EcdR; ECR; EcR-A; EcR-B; EcR-B1; ECR-C; EcR1b; EcRA; EcRB; EcRB-1; EcRB1; EcRC; lie; ms(2)06410; ms(2)42A; NR1H1; snt; USP
UniProt ID: (Fruit fly) P34021
Entrez Gene ID: (Fruit fly) 35540
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.