Novus Biologicals
Manufacturer Code:NBP259251
Catalog # NBP259251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The exact immunogen is proprietary information. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1 EC 2.1.1.43 enhancer of zeste (Drosophila) homolog 2 enhancer of zeste 2 Enhancer of zeste homolog 2 enhancer of zeste homolog 2 (Drosophila) ENX1 ENX-1KMT6EZH1KMT6A histone-lysine N-methyltransferase EZH2 Lysine N-methyltransferase 6 MGC9169; Enhancer of zeste homolog 2; ENX-1; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6
Gene Aliases: ENX-1; ENX1; EZH1; EZH2; EZH2b; KMT6; KMT6A; WVS; WVS2
UniProt ID: (Human) Q15910
Entrez Gene ID: (Human) 2146
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.