OriGene
Manufacturer Code:TA803011
Catalog # 50-167-9539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10B10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 182-431 of human EZH2 ( NP&lowbar004447) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Enhancer of zeste homolog 2; ENX-1; ENX-1 ENX1 EZH2b KMT6 KMT6A WVS WVS2; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6
Gene Aliases: ENX-1; ENX1; EZH1; EZH2; EZH2b; KMT6; KMT6A; WVS; WVS2
UniProt ID: (Human) Q15910
Entrez Gene ID: (Human) 2146
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.