Invitrogen
Manufacturer Code:14986737
Catalog # 14-986-737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AC22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: an enhancer of zeste 2 polycomb repressive complex 2 subunit enhancer of zeste homolog 2 enhancer of zeste homolog 2 (Drosophila) ENX 1 ENX1 Enx-1 Enx1h EZH 2 EZH1 EZH2 EZH-2 EZH2b Histone-lysine N-methyltransferase EZH2 KMT6 KMT6A Lysine N-methyltransferase 6 mKIAA4065 OTTHUMP00000213774 OTTHUMP00000213778 OTTHUMP00000213779 OTTHUMP00000213780 WVS WVS2; Enhancer of zeste homolog 2; ENX-1; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6
Gene Aliases: ENX-1; ENX1; Enx1h; EZH1; EZH2; EZH2b; KMT6; KMT6A; mKIAA4065; WVS; WVS2
UniProt ID: (Human) Q15910, (Mouse) Q61188
Entrez Gene ID: (Human) 2146, (Rat) 312299, (Mouse) 14056
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.