Invitrogen
Manufacturer Code:PA521138
Catalog # PIPA521138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the amino terminus of human EZH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Enhancer of zeste homolog 2; ENX-1; ENX-1 ENX1 EZH1 KMT6 KMT6A enhancer of zeste 2 histone-lysine N-methyltransferase EZH2 lysine N-methyltransferase 6; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6
Gene Aliases: ENX-1; ENX1; Enx1h; EZH1; EZH2; EZH2b; KMT6; KMT6A; mKIAA4065; WVS; WVS2
UniProt ID: (Human) Q15910, (Mouse) Q61188
Entrez Gene ID: (Human) 2146, (Mouse) 14056
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.