Abnova
Manufacturer Code:PAB19619
Catalog # 89-121-147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human EZH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of EZH2.
For Research Use Only
Protein Aliases: Enhancer of zeste homolog 2; ENX-1; EZH1; Histone-lysine N-methyltransferase EZH2; KMT6; Lysine N-methyltransferase 6; MGC9169
Gene Aliases: ENX-1; ENX1; EZH1; EZH2; EZH2b; KMT6; KMT6A; WVS; WVS2
UniProt ID: (Human) Q15910
Entrez Gene ID: (Human) 2146
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.