Invitrogen
Manufacturer Code:PA541428
Catalog # PIPA541428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human EZH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Enhancer of zeste homolog 1; Enhancer of zeste homolog 1 (Drosophila) KIAA0388 KMT6B; ENX-2; Histone-lysine N-methyltransferase EZH1
Gene Aliases: ENX-2; Enx2; EZH1; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829
UniProt ID: (Bovine) A7E2Z2, (Human) Q92800, (Mouse) P70351
Entrez Gene ID: (Bovine) 533087, (Dog) 480516, (Horse) 100052411, (Human) 2145, (Rabbit) 100351852, (Guinea pig) 100720693, (Rat) 303547, (Mouse) 14055, (Zebrafish) 664754
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.