Novus Biologicals
Manufacturer Code:NB10056358SS
Catalog # NB10056358T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 350-400 of human EZH1 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.43 Enhancer of zeste homolog 1 enhancer of zeste homolog 1 (Drosophila) ENX-2 histone-lysine N-methyltransferase EZH1 KIAA0388enhancer of zeste (Drosophila) homolog 1 KMT6B; Enhancer of zeste homolog 1; ENX-2; Histone-lysine N-methyltransferase EZH1
Gene Aliases: EZH1; KIAA0388; KMT6B
UniProt ID: (Human) Q92800
Entrez Gene ID: (Human) 2145
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.