Proteintech
Manufacturer Code:208521AP150UL
Catalog # 50-173-3119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EZH1 Fusion Protein Ag14732 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Enhancer of zeste homolog 1; Enhancer of zeste homolog 1 ENX 2 EZH1 KIAA0388; ENX-2; Histone-lysine N-methyltransferase EZH1
Gene Aliases: ENX-2; Enx2; EZH1; KIAA0388; KMT6B
UniProt ID: (Human) Q92800, (Mouse) P70351
Entrez Gene ID: (Human) 2145, (Mouse) 14055
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.