Invitrogen
Manufacturer Code:PA519220
Catalog # PIPA519220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TDPTAEYSTIHSP) corresponding to the internal amino acids of EYA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EYA1 eyes absent homolog 1 (Drosophila) BOP BOR MGC141875 Eyes absent Drosophila homolog of 1 Melnick-Fraser syndrome OTTHUMP00000195053 eyes absent 1; Eyes absent homolog 1
Gene Aliases: BOP; BOR; BOS1; EYA1; OFC1
UniProt ID: (Human) Q99502
Entrez Gene ID: (Human) 2138
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.