Novus Biologicals
Manufacturer Code:NBP19834720UL
Catalog # NBP19834720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Eya1 - middle region. Peptide sequence SGFNSSQQDYPSYPGFGQGQYAQYYNSSPYPAHYMTSSNTSPTTPSTNAT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOP BOR EC 3.1.3.48 eyes absent (Drosophila) homolog 1 eyes absent homolog 1 eyes absent homolog 1 (Drosophila) MGC141875; Eyes absent homolog 1
Gene Aliases: BOP; BOR; BOS1; EYA1; OFC1
UniProt ID: (Human) Q99502
Entrez Gene ID: (Human) 2138
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.