Invitrogen
Manufacturer Code:PA576156
Catalog # PIPA576156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human EXTL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin tumor-like 3; Exostosin-like 3; exostosin-like glycosyltransferase 3; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Hereditary multiple exostoses gene isolog; Multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; Reg receptor; RPR BOTV REGR EXTR1 EXTL1L
Gene Aliases: 2900009G18Rik; AU040105; BOTV; Ext1l; EXTL1L; EXTL3; EXTR1; KIAA0519; mKIAA0519; REGR; RPR
UniProt ID: (Human) O43909
Entrez Gene ID: (Human) 2137, (Rat) 56819, (Mouse) 54616
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.