Invitrogen
Manufacturer Code:PA5112818
Catalog # PIPA5112818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Exostosin-like 3 protein (2-245AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900009G18Rik AU040105 BOTV exostoses (multiple)-like 3 exostoses-like 3 exostosin exostosin like glycosyltransferase 3 exostosin tumor-like 3 Exostosin-like 3 exostosin-like glycosyltransferase 3 Ext1l EXTL1L EXTL3 EXTR1 EXT-related 1 EXT-related protein 1 Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase Hereditary multiple exostoses gene isolog KIAA0519 mKIAA0519 Multiple exostosis-like protein 3 Putative tumor suppressor protein EXTL3 REG reg receptor Regr RPR; exostoses (multiple)-like 3; exostosin tumor-like 3; Exostosin-like 3; exostosin-like glycosyltransferase 3; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Hereditary multiple exostoses gene isolog; Multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; reg receptor
Gene Aliases: BOTV; EXTL1L; EXTL3; EXTR1; KIAA0519; REGR; RPR
UniProt ID: (Human) O43909
Entrez Gene ID: (Human) 2137
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.