Invitrogen
Manufacturer Code:PA526241
Catalog # PIPA526241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 269-295 amino acids from the C-terminal region of human EXTL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; exostoses (multiple)-like 2; exostoses-like 2; Exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXT-related protein 2; EXTR2 EXT-related protein 2 alpha-14-N-acetylhexosaminyltransferase EXTL2 alpha-14-N-acteylhexosaminyltransferase alpha-GalNAcT EXTL2 exostosin-like 2 glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase processed exostosin-like 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Processed exostosin-like 2
Gene Aliases: 3000001D04Rik; AW146439; EXTL2; EXTR2
UniProt ID: (Human) Q9UBQ6, (Mouse) Q9ES89
Entrez Gene ID: (Human) 2135, (Mouse) 58193
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.