Invitrogen
Manufacturer Code:PA576886
Catalog # PIPA576886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human EXT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: exostoses (multiple) 2; exostosin 2; Exostosin-2; Exostosin-2 Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N acetylglucosaminyl-proteoglycan 4-alpha-N acetylglucosaminyltransferase Multiple exostoses protein 2 Putative tumor suppressor protein EXT2 EXT2; Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Multiple exostoses protein 2; Multiple exostoses protein 2 homolog; N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase; Putative tumor suppressor protein EXT2
Gene Aliases: AI893565; EXT2; SOTV; SSMS
UniProt ID: (Human) Q93063, (Mouse) P70428
Entrez Gene ID: (Human) 2132, (Mouse) 14043, (Rat) 311215
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.