Invitrogen
Manufacturer Code:PA5116952
Catalog # PIPA5116952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 467-746 of human EXT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA409028 exostoses (multiple) 1 exostosin 1 exostosin glycosyltransferase 1 exostosin-1 EXT Ext1 Glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase Langer-Giedion syndrome chromosome region LGCR LGS multiple exostoses protein 1 Multiple exostoses protein 1 homolog N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase putative tumor suppressor protein EXT1 TRPS2 TTV; exostoses (multiple) 1; exostosin 1; Exostosin-1; Glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase; Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Langer-Giedion syndrome chromosome region; Multiple exostoses protein 1; Multiple exostoses protein 1 homolog; N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase; Putative tumor suppressor protein EXT1
Gene Aliases: AA409028; EXT; EXT1; LGCR; LGS; TRPS2; TTV
UniProt ID: (Human) Q16394, (Mouse) P97464
Entrez Gene ID: (Human) 2131, (Mouse) 14042
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.