Invitrogen
Manufacturer Code:PA512378
Catalog # PIPA512378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 243-276 amino acids from the C-terminal region of human EXOSC8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CBP-interacting protein 3; Exosome complex component RRP43; exosome complex exonuclease RRP43; Exosome complex exonuclease RRP43 Ribosomal RNA-processing protein 43 Exosome component 8 p9 Opa-interacting protein 2 EXOSC8 OIP2 RRP43; Exosome component 8; OIP-2; Opa interacting protein 2; Opa-interacting protein 2; p9; Ribosomal RNA-processing protein 43
Gene Aliases: bA421P11.3; CIP3; EAP2; EXOSC8; OIP2; p9; PCH1C; RRP43; Rrp43p
UniProt ID: (Human) Q96B26
Entrez Gene ID: (Human) 11340
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.