OriGene
Manufacturer Code:TA504227
Catalog # 50-167-4955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F5 |
Immunogen | Full length human recombit protein of human EXOSC7 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EAP1 RRP42 Rrp42p hRrp42p p8; Exosome complex component RRP42; exosome complex exonuclease RRP42; Exosome component 7; p8; Ribosomal RNA-processing protein 42
Gene Aliases: EAP1; EXOSC7; hRrp42p; KIAA0116; p8; RRP42; Rrp42p
UniProt ID: (Human) Q15024
Entrez Gene ID: (Human) 23016
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.