Abnova
Manufacturer Code:PAB30061
Catalog # 89-033-349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human EXOSC7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAP1; Exosome complex component RRP42; exosome complex exonuclease RRP42; Exosome component 7; FLJ26543; hRrp42p; KIAA0116; p8; Ribosomal RNA-processing protein 42; RRP42; Rrp42p
Gene Aliases: EAP1; EXOSC7; hRrp42p; KIAA0116; p8; RRP42; Rrp42p
UniProt ID: (Human) Q15024
Entrez Gene ID: (Human) 23016
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.