Invitrogen
Manufacturer Code:PA569113
Catalog # PIPA569113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human EXOSC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exosome complex component MTR3; Exosome complex component MTR3 Exosome component 6 mRNA transport regulator 3 homolog hMtr3 p11 EXOSC6 MTR3; exosome complex exonuclease MTR3; Exosome component 6; hMtr3; homolog of yeast mRNA transport regulator 3; mRNA transport regulator 3; mRNA transport regulator 3 homolog; Mtr3 (mRNA transport regulator 3)-homolog; p11
Gene Aliases: 2610510N21Rik; C76919; EAP4; EXOSC6; hMtr3p; MTR3; Mtr3p; p11
UniProt ID: (Human) Q5RKV6, (Mouse) Q8BTW3
Entrez Gene ID: (Human) 118460, (Bovine) 100126247, (Pig) 100737302, (Mouse) 72544, (Rat) 307850
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.