Abnova
Manufacturer Code:PAB30060
Catalog # 89-033-348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human EXOSC6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAP4; Exosome complex component MTR3; exosome complex exonuclease MTR3; Exosome component 6; hMtr3; hMtr3p; homolog of yeast mRNA transport regulator 3; mRNA transport regulator 3 homolog; MTR3; Mtr3 (mRNA transport regulator 3)-homolog; Mtr3p; p11
Gene Aliases: EAP4; EXOSC6; hMtr3p; MTR3; Mtr3p; p11
UniProt ID: (Human) Q5RKV6
Entrez Gene ID: (Human) 118460
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.