Invitrogen
Manufacturer Code:PA569103
Catalog # PIPA569103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human EXOSC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chronic myelogenous leukemia tumor antigen 28; Exosome complex component RRP46; exosome complex exonuclease RRP46; Exosome component 5; exosome component Rrp46; p12B; Ribosomal RNA-processing protein 46; RRP41B RRP46 Rrp46p hRrp46p p12B chronic myelogenous leukemia tumor antigen 28 exosome complex component RRP46 exosome complex exonuclease RRP46 exosome component Rrp46 ribosomal RNA-processing protein 46
Gene Aliases: CML28; D7Wsu180e; EXOSC5; hRrp46p; p12B; RRP41B; RRP46; Rrp46p; si:ch211-194n6.2; zgc:153707
UniProt ID: (Human) Q9NQT4, (Mouse) Q9CRA8
Entrez Gene ID: (Dog) 611350, (Rabbit) 100357530, (Horse) 100064997, (Human) 56915, (Mouse) 27998, (Rat) 308441, (Guinea pig) 100714897, (Zebrafish) 751632
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.