Invitrogen
Manufacturer Code:PA5109700
Catalog # PIPA5109700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-235 of human EXOSC5 (NP_064543.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chronic myelogenous leukemia tumor antigen 28; Chronic myelogenous leukemia tumor antigen 28 CML28 D7Wsu180e Exosc5 Exosome complex component RRP46 Exosome component 5 hRrp46p p12B Ribosomal RNA-processing protein 46 RRP41B RRP46 Rrp46p si:ch211-194n6.2 zgc:153707; Exosome complex component RRP46; exosome complex exonuclease RRP46; Exosome component 5; exosome component Rrp46; p12B; Ribosomal RNA-processing protein 46
Gene Aliases: CML28; EXOSC5; hRrp46p; p12B; RRP41B; RRP46; Rrp46p
UniProt ID: (Human) Q9NQT4
Entrez Gene ID: (Human) 56915, (Rat) 308441
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.