Invitrogen
Manufacturer Code:PA541890
Catalog # PIPA541890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human EXOSC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exosome complex component RRP41; exosome complex exonuclease RRP41; Exosome component 4; exosome component Rrp41; p12A; ribosomal RNA processing protein 41; Ribosomal RNA-processing protein 41; rrp41
Gene Aliases: 1110039I09Rik; 1500001N04Rik; EXOSC4; hRrp41p; p12A; RRP41; RRP41A; Rrp41p; SKI6; Ski6p; zgc:73175
UniProt ID: (Bovine) Q7YRA3, (Human) Q9NPD3, (Mouse) Q921I9
Entrez Gene ID: (Dog) 482086, (Bovine) 618292, (Human) 54512, (Rabbit) 100342181, (Horse) 100063419, (Mouse) 109075, (Rat) 300045, (Guinea pig) 100712893, (Zebrafish) 393712
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.