Proteintech
Manufacturer Code:148051AP150UL
Catalog # 50-172-9735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EXOSC2 Fusion Protein Ag6525 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EXOSC2 exosome component 2 hRrp4p p7 RRP4 Rrp4p; Exosome complex component RRP4; exosome complex exonuclease RRP4; Exosome component 2; homolog of yeast RRP4 (ribosomal RNA processing 4), 3' 5' exoribonuclease (RRP4); homolog of Yeast RRP4 (ribosomal RNA processing 4), 3'-5'-exoribonuclease; Ribosomal RNA-processing protein 4
Gene Aliases: EXOSC2; hRrp4p; p7; RRP4; Rrp4p
UniProt ID: (Human) Q13868, (Mouse) Q8VBV3
Entrez Gene ID: (Human) 23404, (Mouse) 227715, (Rat) 366017
Molecular Function: RNA binding protein esterase exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.