Invitrogen
Manufacturer Code:PA5100508
Catalog # PIPA5100508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EXOSC10 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: autoantigen PM-SCL; Autoantigen PM/Scl 2; Autoantigen PM/Scl 2 Autoantigen PM/Scl 2 homolog autoantigen PM-SCL EXOSC10 Exosome component 10 P100 polymyositis-scleroderma overlap syndrome-associated autoantigen p2 p3 p4 PM/Scl-100 PMSCL PM-Scl PMSCL2 Polymyositis/scleroderma autoantigen 100 kDa polymyositis/scleroderma autoantigen 2 Polymyositis/scleroderma autoantigen 2 homolog RRP6 Rrp6p; Autoantigen PM/Scl 2 homolog; Exosome component 10; P100 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl-100; Polymyositis/scleroderma autoantigen 100 kDa; Polymyositis/scleroderma autoantigen 2; Polymyositis/scleroderma autoantigen 2 homolog
Gene Aliases: EXOSC10; p2; p3; p4; PM-Scl; PM/Scl-100; PMSCL; PMSCL2; RRP6; Rrp6p
UniProt ID: (Human) Q01780, (Mouse) P56960
Entrez Gene ID: (Human) 5394, (Mouse) 50912, (Rat) 313707
Molecular Function: RNA binding protein exoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.