OriGene
Manufacturer Code:CF808513
Catalog # 50-166-8901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H8 |
Immunogen | Full length human recombit protein of human EXOSC1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3'-5' exoribonuclease CSL4 homolog; CGI-108 CSL4 Csl4p SKI4 Ski4p p13; exosomal core protein CSL4; Exosome complex component CSL4; Exosome component 1; homolog of yeast exosomal core protein CSL4
Gene Aliases: CGI-108; CSL4; Csl4p; EXOSC1; p13; SKI4; Ski4p
UniProt ID: (Human) B2R9B3
Entrez Gene ID: (Human) 51013
Molecular Function:
RNA binding protein
exoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.