Proteintech
Manufacturer Code:125851AP150UL
Catalog # 50-172-8086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EXOSC1 Fusion Protein Ag3286 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-5' exoribonuclease CSL4 homolog; CGI 108 CSL4 Csl4p EXOSC1 exosome component 1 hCsl4p p13 SKI4 Ski4p; exosomal core protein CSL4; Exosome complex component CSL4; Exosome component 1; homolog of yeast exosomal core protein CSL4
Gene Aliases: 2610035C18Rik; 2610104C07Rik; 2610312F07Rik; AI447561; CGI-108; CSL4; Csl4p; EXOSC1; p13; SKI4; Ski4p
UniProt ID: (Human) B2R9B3, (Mouse) Q9DAA6
Entrez Gene ID: (Human) 51013, (Rat) 679140, (Mouse) 66583
Molecular Function:
RNA binding protein
exoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.