Invitrogen
Manufacturer Code:PA571463
Catalog # PIPA571463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 360-389 amino acids from the Central region of human EXOC8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex 84 kDa subunit; Exocyst complex component 8; Exocyst complex component 8 Exocyst complex 84 kDa subunit EXOC8
Gene Aliases: AI414418; EXO84; Exo84p; EXOC8; R74783; SEC84
UniProt ID: (Human) Q8IYI6, (Mouse) Q6PGF7
Entrez Gene ID: (Human) 149371, (Mouse) 102058
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.