Invitrogen
Manufacturer Code:PA554618
Catalog # PIPA554618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human EXOC7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EXOC7 exocyst complex component 7 HGNC:23214 2-5-3p EXO70 Exo70p YJL085W; Exocyst complex component 7; Exocyst complex component Exo70; exocyst component protein 70 kDa homolog; rExo70
Gene Aliases: 2-5-3p; EX070; EXO70; Exo70p; EXOC1; EXOC7; KIAA1067; sec70; YJL085W
UniProt ID: (Human) Q9UPT5, (Mouse) O35250, (Rat) O54922
Entrez Gene ID: (Human) 23265, (Mouse) 53413, (Rat) 64632
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.