Novus Biologicals
Manufacturer Code:NBP299472AF532
Catalog # NB061035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human EXOC5. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 5; exocyst complex component 5 Exocyst complex component Sec10 HSEC10 PRO1912 SEC10 (S. cerevisiae)-like 1 SEC10DKFZp666H126 SEC10L1 SEC10-like 1 SEC10-like 1 (S. cerevisiae) SEC10P; Exocyst complex component Sec10; hSec10; SEC10-like 1
Gene Aliases: EXOC5; HSEC10; PRO1912; SEC10; SEC10L1; SEC10P
UniProt ID: (Human) O00471
Entrez Gene ID: (Human) 10640
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.