Proteintech
Manufacturer Code:175931AP150UL
Catalog # 50-173-1672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EXOC5 Fusion Protein Ag11524 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 71 kDa component of rsec6/8 secretory complex; EXOC5 exocyst complex component 5 HSEC10 PRO1912 SEC10 SEC10L1 SEC10P; Exocyst complex component 5; Exocyst complex component Sec10; hSec10; p71; p71 component of rsec6/8 secretory complex; SEC10-like 1
Gene Aliases: AI447711; AI448003; EXOC5; Gm76; HSEC10; PRO1912; SEC10; SEC10L1; SEC10P
UniProt ID: (Human) O00471, (Rat) P97878, (Mouse) Q3TPX4
Entrez Gene ID: (Human) 10640, (Rat) 60627, (Mouse) 105504
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.