OriGene
Manufacturer Code:TA811243
Catalog # 50-168-3931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7G9 |
Immunogen | Full length human recombit protein of human EXOC4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 4; Exocyst complex component Sec8; SEC8 SEC8L1 Sec8p; SEC8-like 1
Gene Aliases: EXOC4; KIAA1699; SEC8; SEC8L1; Sec8p
UniProt ID: (Human) Q96A65
Entrez Gene ID: (Human) 60412
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.