Proteintech
Manufacturer Code:665371IG150UL
Catalog # 50-173-6958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C1C2 |
Immunogen | EXOC3L2 Fusion Protein Ag26912 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EXOC3L2 XTP7; exocyst complex component 3-like 2; Exocyst complex component 3-like protein 2; HBV X-transactivated gene 7 protein; HBV XAg-transactivated protein 7; protein 7 transactivated by hepatitis B virus X antigen (HBxAg)
Gene Aliases: EXOC3L2; XTP7
UniProt ID: (Human) Q8N9W2
Entrez Gene ID: (Human) 90332
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.