Abnova
Manufacturer Code:PAB23832
Catalog # 89-125-541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human EXOC3L2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant EXOC3L2.
For Research Use Only
Protein Aliases: exocyst complex component 3-like 2; Exocyst complex component 3-like protein 2; FLJ36147 MGC16332 XTP7; HBV X-transactivated gene 7 protein; HBV XAg-transactivated protein 7; protein 7 transactivated by hepatitis B virus X antigen (HBxAg)
Gene Aliases: EXOC3L2; XTP7
UniProt ID: (Human) Q8N9W2
Entrez Gene ID: (Human) 90332
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.