Invitrogen
Manufacturer Code:PA535040
Catalog # PIPA535040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 652-680 amino acids from the C-terminal region of human EXOC3L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EXOC3L1 EXOC3L Exocyst complex component 3-like protein Protein Jiangli; exocyst complex component 3-like 1; Exocyst complex component 3-like protein; Protein Jiangli
Gene Aliases: C730015A04Rik; EXOC3L; EXOC3L1
UniProt ID: (Human) Q86VI1, (Mouse) Q8BI71
Entrez Gene ID: (Human) 283849, (Mouse) 277978
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.