Invitrogen
Manufacturer Code:PA532149
Catalog # PIPA532149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 475 and 752 of Human SEC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 3; Exocyst complex component Sec6; Sec 6 homolog; SEC6 SEC6L1 Sec6p SEC6-like 1 Sec 6 homolog exocyst complex component Sec6; SEC6-like 1
Gene Aliases: 2810050O03Rik; E430013E20Rik; EXOC3; SEC6; SEC6L1; Sec6p
UniProt ID: (Human) O60645, (Mouse) Q6KAR6
Entrez Gene ID: (Human) 11336, (Mouse) 211446
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.