Invitrogen
Manufacturer Code:PA5112887
Catalog # PIPA5112887
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Exocyst complex component 3 protein (410-756AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810050O03Rik BOS_19945 E430013E20Rik Exoc3 Exocyst complex component 3 Exocyst complex component Sec6 I79_024581 LOC100356252 rSec6 Sec 6 homolog Sec6 Sec6 protein SEC6L1 SEC6-like 1 Sec6p; Exocyst complex component 3; Exocyst complex component Sec6; Sec 6 homolog; SEC6-like 1
Gene Aliases: EXOC3; SEC6; SEC6L1; Sec6p
UniProt ID: (Human) O60645
Entrez Gene ID: (Human) 11336
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.