Invitrogen
Manufacturer Code:PA546730
Catalog # PIPA546730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human EXOC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 2; Exocyst complex component 2-like protein; Exocyst complex component Sec5; rSec5; SEC5 homolog; Sec5 protein; SEC5 SEC5L1 Sec5p SEC5-like 1 exocyst complex component Sec5; SEC5-like 1
Gene Aliases: 2410030I24Rik; AI648199; EXOC2; Rsec5; SEC5; SEC5L1; Sec5p
UniProt ID: (Human) Q96KP1, (Mouse) Q9D4H1, (Rat) O54921
Entrez Gene ID: (Human) 55770, (Rabbit) 100354138, (Dog) 478696, (Horse) 100049898, (Bovine) 513431, (Mouse) 66482, (Guinea pig) 100714267, (Rat) 171455
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.