Novus Biologicals
Manufacturer Code:NBP287377
Catalog # NB034999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human EXOC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 2; exocyst complex component 2 Exocyst complex component Sec5 FLJ11026 SEC5 SEC5L1 SEC5-like 1 SEC5-like 1 (S. cerevisiae) Sec5p; Exocyst complex component Sec5; SEC5-like 1
Gene Aliases: EXOC2; SEC5; SEC5L1; Sec5p
UniProt ID: (Human) Q96KP1
Entrez Gene ID: (Human) 55770
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.