Abnova
Manufacturer Code:PAB22609
Catalog # 89-124-321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human EXOC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant EXOC2.
For Research Use Only
Protein Aliases: Exocyst complex component 2; Exocyst complex component Sec5; FLJ11026; SEC5; SEC5-like 1; SEC5L1; Sec5p
Gene Aliases: EXOC2; SEC5; SEC5L1; Sec5p
UniProt ID: (Human) Q96KP1
Entrez Gene ID: (Human) 55770
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.