Invitrogen
Manufacturer Code:PA558071
Catalog # PIPA558071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human EXOC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM-102 SEC3 SEC3L1 SEC3P SEC3-like 1 exocyst complex component Sec3; Exocyst complex component 1; Exocyst complex component Sec3; SEC3-like 1
Gene Aliases: BM-012; BM-102; EXOC1; SEC3; SEC3L1; SEC3P
UniProt ID: (Human) Q9NV70
Entrez Gene ID: (Human) 55763
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.